Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slo-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029356.4 | NP_001024527.1 | 2 - 3 | 497 | 77 | |
NM_001029357.5 | NP_001024528.1 | 2 - 3 | 497 | 77 | ||
NM_001029358.5 | NP_001024529.1 | 2 - 3 | 178 | 77 | ||
NM_001136452.4 | NP_001129924.1 | 2 - 3 | 181 | 77 | ||
NM_001270215.3 | NP_001257144.1 | 4 - 5 | 289 | 77 | ||
NM_001270216.3 | NP_001257145.1 | 3 - 4 | 244 | 77 | ||
NM_001270217.3 | NP_001257146.1 | 3 - 4 | 298 | 77 | ||
NM_001270218.3 | NP_001257147.1 | 3 - 4 | 259 | 77 | ||
GenBank mRNA | AF173828.1 | - | 2 - 3 | 389 | 77 | |
AF232770.1 | - | 2 - 3 | 190 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |