Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270229.4 | NP_001257158.1 | 8 - 9 | 1422 | 114 | |
NM_001270230.4 | NP_001257159.1 | 7 - 8 | 1155 | 114 | ||
NM_001270232.4 | NP_001257161.1 | 2 - 3 | 300 | 114 | ||
NM_001373507.3 | NP_001359540.1 | 25 | 5092 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |