Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077535.7 | NP_509936.3 | 4 - 5 | 561 | 87 | |
GenBank mRNA | AY887904.1 | - | 4 - 5 | 583 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |