Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077541.4 | NP_509942.4 | 8 - 9 | 961 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |