Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077623.3 | NP_510024.2 | 4 - 5 | 414 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |