Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029416.4 | NP_001024587.1 | 1 - 2 | 94 | 125 | |
GenBank mRNA | X92564.1 | - | 78 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |