Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083303.5 | NP_001076772.1 | 13 - 14 | 1687 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |