Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077842.4 | NP_510243.1 | 13 - 14 | 1612 | 164 | |
GenBank mRNA | AB012617.1 | - | 13 - 14 | 1518 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |