Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270300.3 | NP_001257229.1 | 2 - 3 | 233 | 106 | |
NM_001270301.4 | NP_001257230.1 | 1 - 2 | 137 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |