Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077940.3 | NP_510341.2 | 3 - 4 | 277 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |