Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077985.6 | NP_510386.2 | 8 - 9 | 1278 | 126 | |
GenBank mRNA | AB054992.1 | - | 8 - 9 | 1278 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |