Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330918.4 | NP_001317893.1 | 2 - 3 | 94 | 97 | |
NM_001330920.4 | NP_001317903.1 | 1 - 2 | 64 | 97 | ||
NM_001381146.1 | NP_001368607.1 | 2 - 3 | 94 | 97 | ||
NM_001381147.2 | NP_001366850.1 | 2 - 3 | 138 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |