Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gfi-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078008.7 | NP_510409.1 | 6 - 7 | 716 | 98 |
Gene Symbol: M163.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381148.1 | NP_001367098.1 | 6 - 7 | 697 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |