Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-31A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069751.6 | NP_502152.3 | 3 - 4 | 878 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |