Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068036.6 | NP_500437.2 | 5 - 6 | 1034 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |