Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068036.6 | NP_500437.2 | 7 - 8 | 1278 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |