Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062259.6 | NP_494660.2 | 2 | 1480 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |