Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072281.6 | NP_504682.1 | 2 | 1013 | 66 | |
GenBank mRNA | L39895.1 | - | 2 | 1011 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |