Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028399.4 | NP_001023570.2 | 10 - 11 | 1584 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |