Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026805.2 | NP_001021976.1 | 1 - 2 | 192 | 71 | |
GenBank mRNA | DQ015968.1 | - | 1 - 2 | 114 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |