Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027439.2 | NP_001022610.1 | 4 - 5 | 1461 | 121 | |
NM_001382947.1 | 5 - 6 | 1296 | 121 | |||
NM_001382948.1 | 4 - 5 | 1743 | 121 | |||
NR_136383.1 | 4 - 5 | 441 | 121 |
Gene Symbol: F54F2.14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379849.1 | NP_001368592.1 | 5 - 6 | 1199 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |