Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028160.7 | NP_001023331.1 | 1 - 2 | 139 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |