Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640616.3 | XP_005640673.1 | 7 - 8 | 1070 | 62 | |
XM_014111088.2 | XP_013966563.1 | 7 - 8 | 1161 | 62 | ||
XM_022414979.1 | XP_022270687.1 | 8 - 9 | 1265 | 62 | ||
XM_845670.5 | XP_850763.2 | 8 - 9 | 1229 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |