Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003172.2 | NP_001003172.2 | 4 - 5 | 437 | 121 | |
XM_005633274.2 | XP_005633331.1 | 5 - 6 | 600 | 121 | ||
XM_005633278.3 | XP_005633335.1 | 5 - 6 | 578 | 121 | ||
XM_014122414.2 | XP_013977889.1 | 6 - 7 | 796 | 121 | ||
XM_022407418.1 | XP_022263126.1 | 5 - 6 | 1614 | 121 | ||
XM_022407419.1 | XP_022263127.1 | 5 - 6 | 1604 | 121 | ||
XM_022407420.1 | XP_022263128.1 | 5 - 6 | 1550 | 121 | ||
XM_022407421.1 | XP_022263129.1 | 5 - 6 | 1637 | 121 | ||
GenBank mRNA | AB017552.1 | - | 4 - 5 | 437 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |