Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003171.2 | NP_001003171.2 | 3 - 4 | 172 | 56 | |
XM_014111133.2 | XP_013966608.1 | 4 - 5 | 564 | 56 | ||
GenBank mRNA | AB017553.1 | - | 3 - 4 | 172 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |