Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003051.1 | NP_001003051.1 | 1 - 2 | 71 | 52 | |
XM_005617215.3 | XP_005617272.1 | 3 - 4 | 3972 | 52 | ||
GenBank mRNA | AB054642.1 | - | 1 - 2 | 71 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Inventoried Probe spans exons |