Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002949.1 | NP_001002949.1 | 1020 | 85 | ||
GenBank mRNA | AB116145.1 | - | 332 | 85 | ||
AB154172.1 | - | 1020 | 85 | |||
AY509563.1 | - | 579 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Inventoried |