Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031632.1 | NP_001026802.1 | 5 - 6 | 1021 | 86 | |
XM_005628830.3 | XP_005628887.1 | 6 - 7 | 1218 | 86 | ||
XM_014119229.2 | XP_013974704.1 | 6 - 7 | 1217 | 86 | ||
GenBank mRNA | AB183284.2 | - | 5 - 6 | 1021 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |