Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048028.1 | NP_001041493.1 | 6 - 7 | 726 | 113 | |
XM_005633300.3 | XP_005633357.1 | 6 - 7 | 852 | 113 | ||
XM_022407438.1 | XP_022263146.1 | 6 - 7 | 771 | 113 | ||
GenBank mRNA | AB210085.1 | - | 6 - 7 | 726 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |