Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003156.1 | NP_001003156.1 | 2 - 3 | 195 | 82 | |
XM_022409540.1 | XP_022265248.1 | 2 - 3 | 153 | 82 | ||
GenBank mRNA | AF109151.1 | - | 2 - 3 | 195 | 82 | |
AY029186.1 | - | 2 - 3 | 158 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |