Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003135.2 | NP_001003135.2 | 6 - 7 | 1089 | 91 | |
XM_014115499.2 | XP_013970974.2 | 7 - 8 | 3371 | 91 | ||
XM_022420728.1 | XP_022276436.1 | 7 - 8 | 3371 | 91 | ||
XM_022420729.1 | XP_022276437.1 | 7 - 8 | 1355 | 91 | ||
XM_022420730.1 | XP_022276438.1 | 5 - 6 | 2185 | 91 | ||
GenBank mRNA | AF147784.2 | - | 590 | 91 | ||
AF241221.1 | - | 6 - 7 | 1089 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |