Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003116.1 | NP_001003116.1 | 1 - 2 | 238 | 55 | |
GenBank mRNA | AF187325.1 | - | 1 - 2 | 238 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |