Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELANE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003378.2 | NP_001003378.1 | 2 - 3 | 249 | 91 | |
GenBank mRNA | AF494190.1 | - | 2 - 3 | 252 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |