Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536238.6 | XP_536238.1 | 2 | 778 | 77 | |
GenBank mRNA | AJ388534.1 | - | 1 | 481 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |