Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639860.3 | XP_003639908.1 | 6 | 1078 | 128 | |
GenBank mRNA | AJ388540.1 | - | 1 | 352 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |