Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197189.1 | NP_001184118.1 | 1 | 454 | 54 | |
XM_005631027.3 | XP_005631084.1 | 2 | 539 | 54 | ||
GenBank mRNA | AY064408.1 | - | 1 | 268 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |