Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADORA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003279.1 | NP_001003279.1 | 424 | 100 | ||
XM_005622154.3 | XP_005622211.1 | 513 | 100 | |||
XM_005622155.3 | XP_005622212.1 | 472 | 100 | |||
GenBank mRNA | X14051.1 | - | 424 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Canis familiaris |