Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006646.1 | NP_001006647.1 | 465 | 131 | ||
GenBank mRNA | AB716701.1 | - | 423 | 131 | ||
AY572971.1 | - | 465 | 131 | |||
L13027.1 | - | 411 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried |