Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010954.2 | NP_001010954.1 | 3 - 4 | 440 | 64 | |
GenBank mRNA | AY739720.1 | - | 3 - 4 | 441 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |