Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005758.1 | NP_001005758.1 | 2 - 3 | 461 | 97 | |
XM_014114966.2 | XP_013970441.2 | 2 - 3 | 358 | 97 | ||
XM_022420749.1 | XP_022276457.1 | 2 - 3 | 523 | 97 | ||
GenBank mRNA | AY745240.1 | - | 2 - 3 | 461 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |