Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421680.1 | XP_022277388.1 | 396 | 91 | ||
GenBank mRNA | DQ195089.1 | - | 248 | 91 |