Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003286.1 | NP_001003286.1 | 4 - 5 | 502 | 128 | |
GenBank mRNA | X53592.1 | - | 502 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |