Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022407918.1 | XP_022263626.1 | 5 - 6 | 2237 | 125 | |
XM_022407919.1 | XP_022263627.1 | 4 - 5 | 1979 | 125 | ||
XM_534086.5 | XP_534086.3 | 4 - 5 | 2074 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |