Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433296.4 | XP_003433344.1 | 431 | 140 | ||
XM_005635533.3 | XP_005635590.1 | 448 | 140 | |||
XM_005635534.3 | XP_005635591.1 | 448 | 140 | |||
XM_005635535.3 | XP_005635592.1 | 531 | 140 | |||
XM_022409627.1 | XP_022265335.1 | 427 | 140 | |||
XM_022409628.1 | XP_022265336.1 | 444 | 140 | |||
XM_022409629.1 | XP_022265337.1 | 444 | 140 | |||
XM_022409630.1 | XP_022265338.1 | 433 | 140 | |||
XM_534526.6 | XP_534526.2 | 535 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |