Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637593.3 | XP_005637650.1 | 4 - 5 | 439 | 153 | |
XM_005637596.3 | XP_005637653.1 | 2 - 3 | 249 | 153 | ||
XM_014108995.2 | XP_013964470.1 | 4 - 5 | 465 | 153 | ||
XM_014108996.2 | XP_013964471.1 | 4 - 5 | 510 | 153 | ||
XM_014108997.2 | XP_013964472.1 | 5 - 6 | 548 | 153 | ||
XM_014108999.2 | XP_013964474.1 | 3 - 4 | 372 | 153 | ||
XM_022411644.1 | XP_022267352.1 | 2 - 3 | 511 | 153 | ||
XM_022411645.1 | XP_022267353.1 | 5 - 6 | 582 | 153 | ||
XM_022411646.1 | XP_022267354.1 | 5 - 6 | 627 | 153 | ||
XM_022411647.1 | XP_022267355.1 | 5 - 6 | 522 | 153 | ||
XM_022411648.1 | XP_022267356.1 | 4 - 5 | 484 | 153 | ||
XM_534983.5 | XP_534983.2 | 3 - 4 | 346 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |