Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022412897.1 | XP_022268605.1 | 6 - 7 | 808 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |