Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426585.1 | XP_022282293.1 | 801 | 66 | ||
XM_532285.5 | XP_532285.2 | 706 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |