Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616083.3 | XP_005616140.2 | 6 - 7 | 928 | 73 | |
XM_014114388.2 | XP_013969863.1 | 7 - 8 | 1171 | 73 | ||
XM_014114392.2 | XP_013969867.1 | 9 - 10 | 1336 | 73 | ||
XM_022422507.1 | XP_022278215.1 | 7 - 8 | 944 | 73 | ||
XM_022422508.1 | XP_022278216.1 | 9 - 10 | 1109 | 73 | ||
XM_022422509.1 | XP_022278217.1 | 6 - 7 | 697 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |