Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014117326.2 | XP_013972801.1 | 2 - 3 | 206 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |