Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLVRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533666.6 | XP_533666.2 | 2 - 3 | 344 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |